Learn More
AdipoGen Ketanserin . tartrate (50 mg)
SDP
Supplier:  AdipoGen AGCR10014M050
Encompass_Preferred
Chemical. CAS: 83846-83-7. Formula: C22H22FN3O3 . C4H6O6. MW: 395.4 . 150.1. Selective 5-HT2A serotonin receptor antagonist. ATP-sensitive potassium channel inhibitor. hERG potassium channel blocker.
Catalog No. 50-205-3762