Learn More
Medchemexpress LLC Keto-itraconazole | 112560-33-5 | MFCD08063772 | 98.4% | 719.62 g·mol⁻¹ | C35H36Cl2N8O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11778910MG
Encompass_Preferred
Keto-itraconazole is a metabolite of itraconazole and a potent cytochrome P450 3A (CYP3A) inhibitor provided as a research-grade sample for drug metabolism and pharmacology studies.
  • High purity (98.4%) suitable for analytical and pharmacology experiments.
  • Available in a 10 mg quantity for small-scale studies.
  • Molecular formula C35H36Cl2N8O5 and molecular weight 719.62 g·mol⁻¹ for consistent identity.
  • Used as a probe inhibitor or control in CYP3A activity assays.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-28369