Learn More
Medchemexpress LLC Keto Ziprasidone | 884305-07-1 | MFCD30747855 | 99.0% | 426.92 | C21H19ClN4O2S | 5 MG

Supplier: Medchemexpress LLC HY1006485MG
Keto Ziprasidone is a characterized impurity of the antipsychotic ziprasidone supplied for research use only. It is provided as a solid reference standard for analytical, identification, and investigational studies involving serotonin and dopamine receptor antagonists.
- Provides a characterized impurity standard for analytical and identification work.
- CAS number 884305-07-1.
- Molecular formula C21H19ClN4O2S; molecular weight 426.92.
- Purity 98.96% suitable for research applications.
- Solid, off-white to light yellow appearance.
- Storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).
- Available in small milligram pack sizes for reference use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.