Learn More
Medchemexpress LLC (+)-Ketoconazole | 142128-59-4 | 99.8% | C26H28Cl2N4O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0105A50MG
Encompass_Preferred
(+)-Ketoconazole is an imidazole anti-fungal agent and a CYP3A4 inhibitor. It also acts as a cytochrome P450 inhibitor. Clinically, it is indicated for candida infection, dermatophytosis, and folliculitis, but carries toxicity risks. This product is intended for research use only.
  • Imidazole anti-fungal agent
  • CYP3A4 and cytochrome P450 inhibitor
  • Clinically indicated for candida infection, dermatophytosis, and folliculitis
  • Associated with androgen deficiency, liver injuries, teratogenesis, and adrenal gland problems
  • Solid, white to off-white appearance
  • For research use only
Catalog No. 50-004-32299