Learn More
Medchemexpress LLC (+)-Ketoconazole | 142128-59-4 | C26H28Cl2N4O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0105A5MG
Encompass_Preferred
(+)-Ketoconazole is an imidazole anti-fungal agent and a CYP3A4 inhibitor. It has been associated with androgen deficiency features in men treated for chronic mycotic infections and also acts as a cytochrome P450 inhibitor. It has been studied for its potential to affect the antischistosomal properties of quinolines against *Schistosoma mansoni* infection.
  • Targets fungal and cytochrome P450 pathways
  • Inhibits anti-infection and metabolic enzymes
  • Soluble in DMSO (33.33 mg/mL) and various in vivo solvents (≥ 2.5 mg/mL)
  • Clinical indications: candida infection, dermatophytosis, folliculitis
  • Potential toxicity: teratogenesis, liver injuries, adrenal gland problems
  • For research use only
Catalog No. 50-004-32922