Learn More
Cayman Chemical Ketoconazole 1g
SDP
Supplier:  Cayman Chemical 152121 G
Encompass_Preferred
A triazole antifungal agent; has activity against C. albicans, C. krusei, C. tropicalis, C. glabrata, C. parapsilosis, C. neoformans, and A. fumigatus strains (IC50s = 0.031-8 UG/ml); inhibits fungal CYP51; inhibits human CYP3A4 (IC50 = 0.54 µM)
Catalog No. 50-297-0589