Learn More
Selleck Chemical LLC Ketoconazole S1353-1g
SDP
Supplier:  Selleck Chemical LLC S13531G
Encompass_Preferred
Ketoconazole inhibits cyclosporine oxidase and testosterone 6 beta-hydroxylase with IC50 of 0 19 mM and 0 22 mM respectively Ketoconazole is an androgen biosynthesis inhibitor
Catalog No. 50-311-0363