Learn More
Apexbio Technology LLC Ketoconazole 65277-42-1 500mg
SDP
Supplier:  Apexbio Technology LLC A4316500
Encompass_Preferred
Ketoconazole (65277-42-1) is a small-molecule inhibitor targeting cytochrome P450-dependent enzymes including cyclosporine oxidase and testosterone 6 -hydroxylase It is designed to inhibit these catalytic activities thereby modulating the oxidative metabolism of clinically relevant substrates Ketoconazole exerts its biological activity primarily through inhibition of cytochrome P450-mediated pathways In in vitro studies ketoconazole demonstrates inhibitory activity with an IC50 of approximately 0 19 mM for cyclosporine oxidase and about 0 22 mM for testosterone 6 -hydroxylase Based on these pharmacological properties ketoconazole holds research potential in studies of drug-drug interactions hepatic enzyme induction and metabolic profiling assays
Catalog No. 50-000-94025