Learn More
Medchemexpress LLC Ketoconazole | 65277-42-1 | 99.6% | C26H28Cl2N4O4 | 1 G
SDP
Supplier:  Medchemexpress LLC HYB01051G
Encompass_Preferred
Ketoconazole is an imidazole anti-fungal agent and a CYP3A4 and CYP24A1 inhibitor. It has been shown to produce features of androgen deficiency and is a potent exosome biogenesis and/or secretion inhibitor. This product is for research use only.
  • Functions as an imidazole anti-fungal agent
  • Acts as a CYP3A4 and CYP24A1 inhibitor
  • Inhibits hepatic CYP450 and CYT b5 activities
  • Enhances antiproliferative effects
  • Increases systemic calcitriol exposure
  • Promotes activation of caspase-independent apoptosis
  • Potent exosome biogenesis and/or secretion inhibitor
Catalog No. 50-004-32298