Learn More
Abcam Ketoconazole, cytochrome P450 inhibitor, 100MG
Supplier:  Abcam AB120620100MG
Encompass_Preferred
MW 531.4 Da, Purity >99%. Potent cytochrome P450 inhibitor (Ki values are 26.7 and 109 nM at CYP3A4 and CYP3A5, respectively). Antifungal agent. Additionally, shows activity at human androgen receptors.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7414