Learn More
Medchemexpress LLC Ketoconazole (Standard) | 65277-42-1 | C26H28Cl2N4O4 | 5 G
SDP
Supplier:  Medchemexpress LLC HYB01055G
Encompass_Preferred
Ketoconazole is an imidazole anti-fungal agent, functioning as a CYP3A4 and CYP24A1 inhibitor. It has been studied for its role in inhibiting cytochrome P450 and its antischistosomal potential against Schistosoma mansoni infection.
  • Functions as an imidazole anti-fungal agent
  • Inhibits CYP3A4 and CYP24A1
  • Inhibits cytochrome P450
  • Demonstrates antischistosomal potential
  • Enhances antiproliferative effects
  • Increases systemic calcitriol exposure
  • Promotes activation of the caspase-independent apoptosis pathway
  • Potent exosome biogenesis and/or secretion inhibitor
Catalog No. 50-004-32822