Learn More
Medchemexpress LLC Ketoisophorone | 1125-21-9 | MFCD00043119 | 99.9% | 152.19 g/mol | C9H12O2 | 50 G

Supplier: Medchemexpress LLC HY10783250G
Ketoisophorone (4-Oxoisophorone) is a cyclic enone used as a synthesis intermediate and building block in organic chemistry. Supplied for research use, it is commonly employed in the preparation of carotenoids, flavoring agents, and related derivatives, and is provided as a high-purity reagent suitable for laboratory synthesis.
- High purity (99.9%).
- Molecular formula C9H12O2 and molecular weight 152.19 g/mol.
- Useful as a synthesis intermediate and building block for organic synthesis.
- Commonly used in preparation of carotenoids and flavoring agents.
- Available in multiple pack sizes, including 50 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.