Learn More
Cayman Chemical -Ketoprofen-d3 1mg
SDP
Supplier:  Cayman Chemical 281481 MG
Encompass_Preferred
An internal standard for the quantification of (±)-ketoprofen by GC- or LC-MS
Catalog No. 50-293-7837