Learn More
Cayman Chemical -Ketoprofen-d3 500ug
SDP
Supplier:  Cayman Chemical 28148500 UG
Encompass_Preferred
An internal standard for the quantification of (±)-ketoprofen by GC- or LC-MS
Catalog No. 50-294-0867