Learn More
Apexbio Technology LLC Ketorolac 74103-06-3 50mg
SDP
Supplier:  Apexbio Technology LLC B340950
Encompass_Preferred
Ketorolac (CAS 74103-06-3) is a non-steroidal anti-inflammatory compound characterized by its non-selective inhibition of cyclooxygenase (COX) isoenzymes By suppressing COX-mediated conversion of arachidonic acid to prostaglandins ketorolac modulates inflammatory processes pain signaling and fever responses at the molecular level It is widely employed in biomedical research to investigate prostaglandin-dependent pathways and to model the pharmacological consequences of COX inhibition in cellular and in vivo systems
Catalog No. 50-000-96340