Learn More
Medchemexpress LLC Ketorolac | 74103-06-3 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0580100MG
Encompass_Preferred
Ketorolac is a non-steroidal anti-inflammatory drug (NSAID) that functions as a nonselective COX inhibitor. It is utilized in ophthalmic solutions for various conditions and also serves as a DDX3 inhibitor for cancer research. This product is stable under recommended storage conditions and is intended for laboratory use only.
  • Non-steroidal anti-inflammatory drug (NSAID)
  • Nonselective COX inhibitor
  • Used in ophthalmic solutions
  • DDX3 inhibitor for cancer research
  • Stable under recommended storage conditions
  • For research use only
Catalog No. 50-004-09027