Learn More
Medchemexpress LLC Ketorolac | 74103-06-3 | mfcd00887595 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB058050MG
Encompass_Preferred
Ketorolac is a non-steroidal anti-inflammatory drug (NSAID) and a nonselective COX inhibitor. It is used in research for conditions like allergic conjunctivitis, cystoid macular edema, and postoperative ocular inflammation. Additionally, it acts as a DDX3 inhibitor and is applicable in cancer research.
  • Acts as a nonselective COX inhibitor
  • Inhibits COX-1 (IC50 of 20 nM) and COX-2 (IC50 of 120 nM)
  • Used in research for allergic conjunctivitis, cystoid macular edema, intraoperative miosis, and postoperative ocular inflammation and pain
  • Functions as a DDX3 inhibitor for cancer research
  • Effectively kills oral cancer cells in vitro
  • Induces apoptosis and inhibits DDX3 protein expression in H357 cells
  • Demonstrates powerful ocular anti-inflammatory activities in vivo
  • Attenuates damage caused by spinal cord ischemia in rats
  • Reduces oral carcinogenesis in mice
Catalog No. 50-004-08675