Learn More
Selleck Chemical LLC Ketorolac 50mg 74103-06-3
SDP
Supplier:  Selleck Chemical LLC S164650MG
Encompass_Preferred
Ketorolac is a non-selective COX inhibitor of COX-1 and COX-2 with IC50 of 1.23 ?M and 3.50 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-523-1