Learn More
Medchemexpress LLC Ketorolac (Standard) | 74103-06-3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0580R5MG
Encompass_Preferred
Ketorolac (Standard) is the analytical standard for Ketorolac, designed for research and analytical applications. It functions as a non-steroidal anti-inflammatory drug (NSAID) and is a nonselective COX inhibitor.
  • Functions as a nonselective COX inhibitor with IC50 values of 20 nM for COX-1 and 120 nM for COX-2.
  • Used as a 0.5% ophthalmic solution for researching allergic conjunctivitis, cystoid macular edema, intraoperative miosis, and postoperative ocular inflammation and pain.
  • Identified as a DDX3 inhibitor with potential for cancer research.
Catalog No. 50-004-09492