Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ketorolac (standard) | 74103-06-3 | MFCD00864281 | 99.7% | 255.27 | C15H13NO3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYB0580R10MG

Encompass_Preferred

Ketorolac (Standard) is an analytical reference standard supplied as a dry solid for research and analytical applications. It is intended for laboratory use and assay development and is not validated for medical or clinical use. Typical specifications include high purity and recommended cold storage.

  • Analytical reference standard for research and assay development.
  • Purity approximately 99.7% (HPLC) as reported on COA.
  • Molecular weight 255.27; formula C15H13NO3; CAS 74103-06-3.
  • Available in small package sizes such as 1 mg, 5 mg, and 10 mg.
  • Recommended storage 4°C sealed and dry; in solution: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-97366


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.