Learn More
Apexbio Technology LLC Ketorolac tromethamine salt 74103-07-4 1g
SDP
Supplier:  Apexbio Technology LLC B14471000
Encompass_Preferred
Ketorolac tromethamine salt (CAS 74103-07-4) is a small-molecule inhibitor that targets cyclooxygenase (COX) enzymes non-selectively resulting in potent analgesic activity through inhibition of prostaglandin synthesis This compound is commonly utilized in biomedical research to investigate pathways related to inflammation pain perception and prostaglandin-mediated physiological responses
Catalog No. 50-000-61999