Learn More
Selleck Chemical LLC Ketorolac tromethamine salt
SDP
Supplier:  Selleck Chemical LLC S56981G
Encompass_Preferred
Ketorolac tromethamine is a synthetic pyrrolizine carboxylic acid derivative with anti-inflammatory analgesic and antipyretic properties It is a non-selective COX inhibitor with IC50s of 20 nM for both COX-1 and COX-2
Catalog No. 50-000-03702