Learn More
Medchemexpress LLC Ketorolac (tromethamine salt) | 74103-07-4 | MFCD00887595 | 99.5% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB01385G
Encompass_Preferred
Ketorolac tromethamine salt is a non-steroidal anti-inflammatory agent, acting as a nonselective COX inhibitor, with IC50s of 20 nM for COX-1 and 120 nM for COX-2. This product is for research use only and is not sold to patients.
  • Non-steroidal anti-inflammatory agent.
  • Nonselective COX inhibitor.
  • Has IC50s of 20 nM for COX-1 and 120 nM for COX-2.
Catalog No. 50-004-50886