Learn More
Medchemexpress LLC Ketotifen | 34580-13-7 | 99.7% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB015750MG
Encompass_Preferred
Ketotifen is an orally active, second-generation noncompetitive histamine 1 (H1) receptor blocker and mast cell stabilizer. It can block 6-phosphogluconate dehydrogenase (PGD) in vitro. Additionally, it exhibits antiviral activity against SARS-CoV-2 and Influenza virus. It is used in the research of autoimmune encephalomyelitis (EAE) and for asthma attack prevention.
  • Orally active, second-generation noncompetitive histamine 1 (H1) receptor blocker
  • Mast cell stabilizer
  • Blocks 6-phosphogluconate dehydrogenase (PGD) in vitro
  • Antiviral activity against SARS-CoV-2 and Influenza virus
  • Used in research of autoimmune encephalomyelitis (EAE)
  • Used for asthma attack prevention
Catalog No. 50-004-06222