Learn More
Palmero Health Care Keyboard Film
Supplier:  Palmero Health Care 1802
Encompass_Preferred
Perforated, plastic sheets with a light adhesive that covers virtually any keyboard. Prevents cross-contamination and leaves behind no residue, does not contain latex. Size: 9" x 19"
Catalog No. NC1632702