Learn More
Medchemexpress LLC KGDS | 93674-95-4 | >99.8% | 405.40 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP35231MG
Encompass_Preferred
KGDS is a synthetic peptide targeting integrin GPIIb-IIIa, found on human activated platelets. Its amino acid sequence is Lys-Gly-Asp-Ser. This product is for research use only and is not intended for patient use.
  • Targets integrin GPIIb-IIIa
  • Amino acid sequence is Lys-Gly-Asp-Ser
  • Intended for research use only
Catalog No. 50-003-96777