Learn More
CPC Scientific H-His-Ser-Gly-Ile-Asn-Ser-Ser-Asn-Ala-Glu-Val-Leu-Ala-Leu-Phe-Asn-Val-Thr-Glu-Met-Asp-Ala-Gly-Glu-Tyr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific KGF001B
Encompass
KGF receptor peptide, which corresponds to residues 199-223 of the KGF receptor alternative exon, blocks the mitogenic activity of KGF and the interaction between KGF and its receptor. The peptide binds directly and specifically to KGF and thus represents a critical part of the KGF receptor ligand binding site.ONE-LETTER SEQUENCE: HSGINSSNAEVLALFNVTEMDAGEYMOLECULAR FORMULA: C114H174N30O42S1MOLECULAR WEIGHT:2668.88STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [161338-76-7], RESEARCH AREA: CancerREFERENCES: D.P.Bottaro et al., J. Biol. Chem., 268, 9180 (1993)
Catalog No. 50-210-9615