Learn More
Medchemexpress LLC KH-4-43 | 2813310-07-3 | 99.7% | 422.74 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14860210MM/1ML
Encompass_Preferred
KH-4-43 is an inhibitor of E3 CRL4, inhibiting its core ligase complex and demonstrating anticancer activity. It is suitable for research use only.
  • Inhibits E3 CRL4 core ligase complex
  • Demonstrates anticancer activity
  • Binding Kd to E3 ROC1-CUL4A CTD is 83 nM
  • Binding Kd to ROC1-CUL1 CTD is 9.4 μM
Catalog No. 50-003-62095