Learn More
Medchemexpress LLC Khasianine | 32449-98-2 | 99.1% | 721.92 g/mol | C39H63NO11 | 10 MG

Supplier: Medchemexpress LLC HYN232210MG
Khasianine is a steroidal glycoalkaloid isolated from black nightshade (Solanum nigrum L.) that has demonstrated antitumor activity in published studies. It is provided as a high-purity research reagent and analytical standard for natural-product and pharmacological research.
- High purity (99.11%).
- CAS number 32449-98-2 for unambiguous identification.
- Molecular formula C39H63NO11 and molecular weight 721.92 g/mol.
- Supplied as a solid suitable for analytical and biological research applications.
- Available in small milligram pack sizes for standard reference use.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.