Learn More
Bon Opus Biosciences Recombinant Human KHDRBS3 Protein(C-His) - Source: E. coli
SDP
Supplier:  Bon Opus Biosciences BP256750UG
Encompass_Preferred
Recombinant Human KHDRBS3 Protein is produced by E.coli expression system and the target gene encoding A11-D300 is expressed with a His tag at the C-terminus. Accession#O75525. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4
Catalog No. NC2733740