Learn More
Medchemexpress LLC Khellin | 82-02-0 | 99.7% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1394100MG
Encompass_Preferred
Khellin is a furochromone isolated from *Ammi visnuga L.* It acts as an EGFR inhibitor with an IC50 of 0.15 μM. Khellin demonstrates anti-proliferative activity *in vitro* and possesses antispasmodic and coronary vasodilator effects.
  • EGFR inhibitor with an IC50 of 0.15 μM.
  • Exhibits anti-proliferative activity in MCF-7 cells and Hela cells.
  • Possesses antispasmodic and coronary vasodilator effects.
  • Inhibits Ca2+ influx pathways activated by noradrenaline and high K+.
Catalog No. 50-004-12525