Learn More
Medchemexpress LLC Khellin | 82-02-0 | 99.7% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1394
Encompass_Preferred
Khellin is a furochromone isolated from *Ammi visnuga L.* that functions as an EGFR inhibitor with an IC50 of 0.15 μM. It exhibits anti-proliferative activity in vitro and possesses antispasmodic and coronary vasodilator effects.
  • Acts as an EGFR inhibitor
  • Demonstrates anti-proliferative activity in vitro
  • Possesses antispasmodic and coronary vasodilator effects
  • Inhibits Ca2+ influx pathways
  • Soluble in DMSO at 16.67 mg/mL
Catalog No. 50-004-14025