Learn More
Medchemexpress LLC HY-10996A 10mg Medchemexpress, KHS101 hydrochloride CAS:1784282-12-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10996A10MG
Encompass_Preferred
Medchemexpress, HY-10996A 10mg KHS101 HCl CAS:1784282-12-7 KHS101 hydrochloride could selectively induce a neuronal differentiation phenotype and interacts with transforming acidic coiled-coil-containing protein 3 (TACC3). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8728