Learn More
Selleck Chemical LLC KHS101 hydrochloride
SDP
Supplier:  Selleck Chemical LLC S591925MG
Encompass_Preferred
KHS101 is a small-molecule inhibitor of TACC3 which is a key component of centrosome-microtubule dynamic networks
Catalog No. 50-000-03937