Learn More
Hach Company Potassium Iodide Swiftest™ Bulk Dispenser, with Potassium Iodide Refill Vial
Supplier:  Hach Company 2834100
Encompass
For total chlorine determination with the Autocat 9000 Chlorine Amperometric Titrator. Includes Swiftest dispenser and vial containing enough reagent for approximately 300 tests.
Catalog No. 50-199-6743