Learn More
Medchemexpress LLC Ki-tox-a3 | 1351116-34-1 | 98.9% | 10 MG

Supplier: Medchemexpress LLC HY17624410MG
KI-TOX-A3 is a selective protein-protein interaction inhibitor that targets the TOX protein. It disrupts the TOX-KAT7 interaction (IC50 = 0.51 μM), promotes proteasomal degradation of TOX, and restores KAT7-mediated H3K14 acetylation. These actions reverse CD8+ T cell exhaustion and inhibit proliferation of T-cell acute lymphoblastic leukemia cells. Supplied as a solid powder for research use.
- Selective PPI inhibitor that targets TOX protein.
- Potent disruption of TOX-KAT7 interaction (IC50 0.51 μM).
- Induces proteasomal degradation of TOX to restore histone acetylation.
- Reverses CD8+ T cell exhaustion in preclinical studies.
- Inhibits proliferation of T-ALL cells.
- High purity (98.9%) and supplied as solid powder.
- Storage: powder -20°C; solutions -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.