Learn More
Medchemexpress LLC Ki-tox-a3 | 1351116-34-1 | >99.15% | 319.31 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY176244
Encompass_Preferred
KI-TOX-A3 is a selective protein-protein interaction (PPI) inhibitor targeting the TOX protein, with an IC50 value of 0.51 μM for TOX-KAT7 interaction. It induces proteasomal degradation of TOX, which restores KAT7-mediated H3K14 acetylation. This action reverses CD8+ T cell exhaustion and inhibits the proliferation of T-cell acute lymphoblastic leukemia (T-ALL) cells. KI-TOX-A3 shows promise for research in hematological tumors such as T-ALL.
  • Selective PPI inhibitor targeting TOX protein
  • Induces proteasomal degradation of TOX
  • Restores KAT7-mediated H3K14 acetylation
  • Reverses CD8+ T cell exhaustion
  • Inhibits T-cell acute lymphoblastic leukemia (T-ALL) cell proliferation
  • Promising for research of hematological tumors
Catalog No. 50-003-84359