Learn More
Medchemexpress LLC KI-TOX-A3 | 1351116-34-1 | 99.15% | 319.31 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1762445MG
Encompass_Preferred
KI-TOX-A3 is a selective protein-protein interaction (PPI) inhibitor that targets the TOX protein. It has an IC50 value of 0.51 μM for the TOX-KAT7 interaction. This compound induces proteasomal degradation of TOX, which restores KAT7-mediated H3K14 acetylation, thereby reversing CD8+ T cell exhaustion and inhibiting the proliferation of T-cell acute lymphoblastic leukemia (T-ALL) cells. KI-TOX-A3 shows promise for research into hematological tumors like T-ALL. It is intended for research use only and not for sale to patients.
  • Selective protein-protein interaction (PPI) inhibitor targeting TOX protein.
  • Induces proteasomal degradation of TOX.
  • Restores KAT7-mediated H3K14 acetylation.
  • Reverses CD8+ T cell exhaustion.
  • Inhibits T-cell acute lymphoblastic leukemia (T-ALL) cell proliferation.
  • Promising for research of hematological tumors (e.g., T-ALL).
Catalog No. 50-003-87290