Learn More
Genetex Inc Ki67 Monoclonal Antibody [MSVA-267M] HistoMAX™ (GTX04361) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX04361
Encompass_Preferred
"The Ki67 antibody [MSVA-267M] HistoMAX™ (GTX04361) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human liver tissue using GTX04361 Ki67 antibody [MSVA-267M] HistoMAX. The liver represents an ideal normal tissue control for Ki67 staining quality. Less than 1 of the hepatocytes should stain and cytoplasmatic staining should be absent."
Catalog No. NC3660441