Learn More
Selleck Chemical LLC Ki8751 5mg 228559-41-9
SDP
Supplier:  Selleck Chemical LLC S13635MG
Encompass_Preferred
Ki8751 is a potent and selective inhibitor of VEGFR2 with IC50 of 0.9 nM, >40-fold selective for VEGFR2 than c-Kit, PDGFR? and FGFR-2, little activity to EGFR, HGFR and InsR. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-712-7