Learn More
Medchemexpress LLC KIF18A-IN-6 | 2914879-10-8 | C28H37N3O5S2 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY153065
Encompass_Preferred
KIF18A-IN-6 (Compound 134) is an orally active KIF18A inhibitor with an IC50 of 0.016 μM against KIF18A microtubule-dependent ATPase activity. It demonstrates anti-proliferative effects on JIMT-1, HCC-15, and NIH-OVCAR3 cells in vitro and exhibits significant tumor growth inhibition in HCC15 and OVCAR3 xenograft models in mice. This product is for research use only.
  • Orally active KIF18A inhibitor
  • High potency against KIF18A microtubule-dependent ATPase activity (IC50 of 0.016 μM)
  • Demonstrates anti-proliferative effects on cells in vitro
  • Exhibits significant tumor growth inhibition in xenograft models in mice
  • Multiple packaging options cater to different research needs
  • Comprehensive documentation ensures quality assurance and provides essential information for researchers
Catalog No. 50-003-74587