Learn More
ABclonal Technology KIF20B Rabbit pAb
SDP
Supplier:  ABclonal Technology A153601000UL
Encompass_Preferred
Enables several functions, including WW domain binding activity, plus-end-directed microtubule motor activity, and protein homodimerization activity. Involved in positive regulation of cell population proliferation and positive regulation of cytokinesis. Located in several cellular components, including microtubule cytoskeleton, midbody, and nuclear lumen.
Catalog No. 50-001-05134