Learn More
Medchemexpress LLC KIF2C-IN-1 (compound 7S9) | 98.1% | 739.23 g/mol | C36H39ClN4O9S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17352210MG
Encompass_Preferred
KIF2C-IN-1 is a selective small-molecule inhibitor that stabilizes the KIF2C-tubulin interaction, blocking microtubule depolymerization and reversing cross-resistance to microtubule-targeting agents. Supplied as a light yellow solid, it has demonstrated cytotoxic activity in paclitaxel-resistant triple-negative breast cancer models and is intended for research use in biological assays.
  • Selective KIF2C inhibition stabilizes microtubules.
  • Demonstrated cytotoxicity in paclitaxel-resistant triple-negative breast cancer models.
  • Fluorescent properties enable visualization (Ex/Em 410/510 nm).
  • High purity suitable for in vitro and in vivo assays.
  • Provided as a small research pack for assay development.
  • Store sealed at 4°C; in solvent store long term at -80°C.
Catalog No. 50-002-95085