Learn More
Eurofins Discoverx KILR Jurkat LAG3 Cell Line, 2 vials. (97-1038C019)
Supplier:  Eurofins Discoverx 971038C019
Encompass
The KILR Jurkat LAG3 cell line is a stable, engineered target cell line to measure direct cell death mediated by immunooncology treatments (e.g. checkpoint inhibitors, checkpoint agonists, bispecifics, CAR-T, and more) via Antibody Dependent Cell-mediated Cytotoxicity (ADCC), Complement Dependent Cytotoxicity (CDC), and Antibody Dependent Cell Phagocytosis (ADCP). This product uses EFC technology. The cell line overexpresses a ProLabel- tagged housekeeping gene as well as untagged LAG3. Upon effector cells-mediated cell lysis, ProLabel is released to the media, where EA in the detection kit produces an EFC signal proportional to the level of ProLabel released. This allows for calculation of percent lysis of the KILR cell line. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kilr.
Catalog No. 50-200-4707
Add to Cart
Request a Quote