Learn More
Crystal Chem Inc Kidney Injury Molecule-1 (KIM-1) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80723
Encompass
The KIM-1 ELISA is a highly sensitive assay that is used to quantify levels of human KIM-1 in plasma, serum, and urine. Highlights: Fast test results in just 3 hours. Reliable and accurate: CV's < 20. Dynamic range of 0 to 4 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7843