Learn More
Enzo Life Sciences KIM-1 (human) ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI9002260001
Encompass_Preferred
Ultra sensitive (1.2 pg/ml) ELISA kit, enabling reduced input sample and matrix interference, for the quantification of KIM-1, an early biomarker for kidney injury or disease. The KIM-1 (human) ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of human KIM-1 in urine with results in <2 hours. Alternative Name: Kidney injury molecule-1. T-cell immunoglobulin and mucin domain-containing protein 1. TIMD-1. TIM-1. HAVCR1. Hepatitis A virus cellular receptor 1. T-cell immunoglobulin mucin receptor 1. CD365. Sensitivity: 1.279 pg/ml (range 7.813 - 500 pg/ml). Assay Time: <2 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human. Contents: Microtiter Plate, Assay Buffer, Standard, Antibody-conjugate, Streptavidin-HRP, TMB Substrate, Stop Solution, Wash Buffer Concentrate
Catalog No. NC0612062