Learn More
Enterprise Technology Solutions Kimberly-Clark Professional Hand Sanitizer, Moisturizing, Foam, 1.2L, 2/CT, CL
SDP
Supplier:  Enterprise Technology Solutions KCC34643
Encompass_Preferred
Show you care by providing Kleenex Ultra Moisturizing Foam Hand Sanitizer in your facility. Kleenex is one of the world's most-loved restroom brands with more than 95 years of experience keeping your staff and guests healthy. This EcoLogo certified solution was formulated to meet the CDC guidelines for alcohol-based sanitizers and helps stop the spread of germs by effectively killing up to 99.9 percent of bacteria - all without the use of water. Hand sanitizer features a luxurious formulation enriched with cucumber and aloe vera so it is gentle for frequent use. It is a popular choice for hotels, spas and more. Because the sanitizer is NSF E-3 rated, it is also suitable for use in food-processing and foodservice settings. Refill is compatible with Kimberly-Clark Professional ICON and Scott Pro Automatic Sanitizer Dispensers (not included).
Catalog No. 50-008-30854