Learn More
Selleck Chemical LLC KIN1148 S0289-5MG
SDP
Supplier:  Selleck Chemical LLC S02895MG
Encompass_Preferred
KIN1148 is an agonist of interferon regulatory factor 3 (IRF3) that induces dose-dependent IRF3 nuclear translocation and specific activation of IRF3-responsive promoters KIN1148 is an influenza vaccine adjuvant that enhances flu vaccine efficacy
Catalog No. 50-000-41111