Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kinase, Natto fermentation | 133876-92-3 | 27728 Da | 10 MG
SDP

Supplier:  Medchemexpress LLC HYP237310MG

Encompass_Preferred

Kinase, Natto fermentation is a potent fibrinolytic enzyme that breaks down blood clots by directly hydrolyzing fibrin and plasmin substrate. It is primarily used for cardiovascular disease research.

  • Breaks down blood clots by directly hydrolyzing fibrin and plasmin substrate.
  • Dissolves 94% of blood clots within ten minutes in vitro.
  • Degrades fibrin clots after two hours of incubation in vitro.
  • Exhibits fibrinolytic activity and dissolves blood clots in vivo.
  • Stable in neutral or alkaline environments (pH 7.0-12.0).
  • Stability improved when mixed with boiled rice extract, broth, serum protein, or gastric juice protein.
  • Retains enzyme activity under acidic conditions with stabilizing agents.
  • Better stability below 37°C.
  • Retains 95% of original activity after 5 freeze-thaw cycles.

Catalog No. 50-004-23461


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.