Learn More
ATCC Kingella kingae; 2941
Supplier:  ATCC 23331
Encompass
Kingella kingae strain 2941 [NCTC 10745] was isolated from blood. This bacterium requires an atmosphere of 5% CO2 for growth.
Catalog No. 50-238-1442